missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAALC Polyclonal Antibody
GREENER_CHOICE

Product Code. 17109553
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17109553 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17109553 Supplier Invitrogen™ Supplier No. PA5111632

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RYYESWTRET ESTWLTYTDS DAPPSAAAPD SGPEAGGLHS VLEAEKSKIK APTDSVSDEG LFSASKMAPL AVFS Highest antigen sequence indentity to the following orthologs: Mouse - 31%, Rat - 53%.

This gene was identified by gene expression studies in patients with acute myeloid leukemia. The gene is conserved among mammals and is not found in lower organisms. Tissues that express this gene develop from the neuroectoderm. Multiple alternatively spliced transcript variants that encode different proteins have been described for this gene; however, some of the transcript variants are found only in AML cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAALC
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BAALC
Gene Accession No. Q8WXS3
Gene Alias 2810457D07Rik; Baalc; Brain and acute leukemia cytoplasmic protein; brain and acute leukemia, cytoplasmic; FLJ12015; HGNC:14333
Gene Symbols BAALC
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human BAALC. Recombinant protein control fragment (Product #RP-107628).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79870
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.