missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAAT1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 462.00 €
Specifications
| Antigen | BAAT1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
BAAT1 Polyclonal antibody specifically detects BAAT1 in Human, Rat samples. It is validated for Western BlotSpecifications
| BAAT1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, DNA Repair | |
| PBS with 50% glycerol, pH7.3. | |
| 221927 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| BAAT1, BRCA1-associated ATM activator 1, BRCA1-associated protein required for ATM activation protein 1, C7orf27, chromosome 7 open reading frame 27, HEAT repeat-containing protein C7orf27, MGC22916 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 652-821 of human BRAT1 (NP_689956.2). AQGLELALVFLGQTLGPPRTHCPYAVALPEVAPAQPLTEALRALCHVGLFDFAFCALFDCDRPVAQKSCDLLLFLRDKIASYSSLREARGSPNTASAEATLPRWRAGEQAQPPGDQEPEAVLAMLRSLDLEGLRSTLAESSDHVEKSPQSLLQDMLATGGFLQGDEADCY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title