missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAAT1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92706-0.1ml
This item is not returnable.
View return policy
Description
BAAT1 Polyclonal antibody specifically detects BAAT1 in Human, Rat samples. It is validated for Western Blot
Specifications
| BAAT1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BAAT1, BRCA1-associated ATM activator 1, BRCA1-associated protein required for ATM activation protein 1, C7orf27, chromosome 7 open reading frame 27, HEAT repeat-containing protein C7orf27, MGC22916 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 652-821 of human BRAT1 (NP_689956.2). AQGLELALVFLGQTLGPPRTHCPYAVALPEVAPAQPLTEALRALCHVGLFDFAFCALFDCDRPVAQKSCDLLLFLRDKIASYSSLREARGSPNTASAEATLPRWRAGEQAQPPGDQEPEAVLAMLRSLDLEGLRSTLAESSDHVEKSPQSLLQDMLATGGFLQGDEADCY | |
| 0.1 mL | |
| Breast Cancer, Cancer, DNA Repair | |
| 221927 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction