missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACH2 Antibody (PCRP-BACH2-5B11), Novus Biologicals™
Mouse Monoclonal Antibody
256.00 € - 484.00 €
Specifications
| Antigen | BACH2 |
|---|---|
| Clone | PCRP-BACH2-5B11 |
| Concentration | 0.2 mg/ml |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30293594
|
Novus Biologicals
NBP3-44619-20ug |
20 μg |
256.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30305451
|
Novus Biologicals
NBP3-44619-100ug |
100 μg |
484.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BACH2 Monoclonal antibody specifically detects BACH2 in Human samples. It is validated for Flow CytometrySpecifications
| BACH2 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 60468 | |
| IgG2a κ | |
| Protein A purified |
| PCRP-BACH2-5B11 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| BTB and CNC homolog 2, BTB and CNC homology 1, basic leucine zipper transcription factor 2, transcription regulator protein BACH2 | |
| Recombinant fragment (around aa11-132) of human BACH2 protein (Uniprot: Q9BYV9). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title