Learn More
Melford Bacitracin
Description
Bacitracin is an antibiotic that has significant importance in the life science marketplace.
In the life science marketplace, bacitracin is primarily used for its antimicrobial properties. It is commonly employed in microbiology laboratories, research, and pharmaceutical industries. Bacitracin is effective against a wide range of Gram-positive bacteria, including Staphylococcus and Streptococcus species.
One of the key applications of bacitracin is in cell culture and tissue culture work. It is used as an additive in cell culture media to prevent bacterial contamination, ensuring the growth of uncontaminated cell lines. Bacitracin acts by inhibiting the synthesis of the bacterial cell wall, leading to the disruption of bacterial growth and preventing contamination.
Moreover, bacitracin is utilized in research laboratories for studying bacterial susceptibility to antibiotics and determining the minimum inhibitory concentrations (MIC) of bacteria. It is an essential tool in evaluating the effectiveness of antimicrobial agents and studying antibiotic resistance mechanisms.
Specifications
Specifications
| Chemical Name or Material | Bacitracin |
| CAS | 1405-87-4 |
| Quantity | 25 g |
| Molecular Formula | C66H103N17O16S |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.