missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bacitracin, potency min. 60 Units/mg

Product Code. 10667542 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10667542 25 g Glass Bottle 25g
10523201 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10667542 Supplier Thermo Scientific Chemicals Supplier No. 226100250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bacitracin, CAS # 1405-87-4, is cyclic polypeptide-based antibiotic which exhibits activity against microbes causing pneumonia, skin, and eye infections.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Bacitracin is a mixture of at least nine different cyclic polypeptides, originally isolated from Bacillus bacteria in 1945

• Bacitracins work by binding divalent metal cations and preventing the hydrolysis of lipids into usable cell wall components

Applications

• Bacitracin has been paired with nanoparticles in some laboratory studies to increase its effectivity

• It is suitable for the laboratory study of bacterial resistance, infection, and cross-reactivity

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-87-4
Molecular Formula C66H103N17O16S
MDL Number MFCD00062640

Specifications

Type Bacitracin
Physical Form Powder
Melting Point 221.0°C to 225.0°C
Infrared Spectrum Authentic
Loss on Drying 5.0% max. (1 g, 60°C, 3 Hrs) (P2O5) (not exceeding 0.1 kPa)
Packaging Glass Bottle
Color Beige or White
Concentration ≥60 I.U./mg (potency, on dried subst.)
Quantity 25 g
Merck Index 15, 927
Solubility Information Solubility in water: soluble. Other solubilities: soluble in glacial acetic acid, methanol and, alcohol, practically insoluble in ether, chloroform and, acetone
Formula Weight 1422.71
Percent Purity 60 I.U./mg min. (potency, on dried subst.)
Chemical Name or Material Bacitracin
Show More Show Less
compliance-icons
Product Identifier
  • Bacitracin
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.