missing translation for 'onlineSavingsMsg'
Learn More

Bacitracin

Product Code. 15414819
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414819 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15414819 Supplier Thermo Scientific Alfa Aesar Supplier No. J62432.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bacitracin, CAS # 1405-87-4, is cyclic polypeptide-based antibiotic which exhibits activity against microbes causing pneumonia, skin, and eye infections. | CAS: 1405-87-4 | C66H103N17O16S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Bacitracin is a mixture of at least nine different cyclic polypeptides, originally isolated from Bacillus bacteria in 1945
  • Bacitracins work by binding divalent metal cations and preventing the hydrolysis of lipids into usable cell wall components

Applications

  • Bacitracin has been paired with nanoparticles in some laboratory studies to increase its effectivity
  • It is suitable for the laboratory study of bacterial resistance, infection, and cross-reactivity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-87-4
Molecular Formula C66H103N17O16S
MDL Number MFCD00062640

Specifications

Type Bacitracin
Physical Form Powder
Melting Point 222°C
Color White
Quantity 25 g
Merck Index 14,934
Solubility Information Soluble in water.
Formula Weight 1421.6
Chemical Name or Material Bacitracin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.