missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Background Plate, 96-well

Product Code. 15784067 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 x 96 well plate
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15784067

Brand: Applied Biosystems™ 4481309

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used to maintain your real-time PCR instrument with 96-well standard block

  • The 96-Well Background Plate is used to maintain your real-time PCR instrument with 96-well standard block.
  • Background calibration should be run at least every six months.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at –20°C ± 5°C.

Packaging 1 x 96 Plate
Product Type Background plate
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
No. of Wells 96
Quantity 1 x 96 well plate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.