Learn More
Fisherbrand™ Bacti-Loop Microsterilizer Loop Holder Attachment
Product Code. 31781852 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (English) Outside:
0.77 in.
Unit Size:
Each
Product Code. Diameter (English) Outside Unit Size
31781852 0.77 in. Each
1 options
Product Code. 31781852
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Helps to support and guide inoculation loops and tools in the Bacti-Loop Microsterilizer.
  • Supports inoculating loops and tools inside the chamber
  • Clamps on to Bacti-Loop body
Specifications
Diameter (English) Outside 0.77 in.
For Use With (Equipment) Bacti-Loop Microsterilizer
Product Type Loop Holder Attachment
Sterility Non-sterile