missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bad, p Ser112 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81807-0.025ML
This item is not returnable.
View return policy
Description
Bad Polyclonal specifically detects Bad in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Bad (p Ser112) | |
| Polyclonal | |
| Unconjugated | |
| phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide | |
| BBC2, BBC6, bcl2 antagonist of cell death, BCL2-antagonist of cell death protein, BCL2-associated agonist of cell death, Bcl-2-binding component 6, BCL2-binding component 6, BCL2-binding protein, Bcl2-L-8, BCL2L8bcl2-L-8, Bcl-2-like protein 8, BCL-X/BCL-2 binding protein, Bcl-XL/Bcl-2-associated death promoter | |
| Rabbit | |
| 23 kDa | |
| 0.025 mL | |
| Phospho Specific | |
| 572 | |
| Primate, Mouse, Human | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Q92934 | |
| BAD | |
| The antiserum was produced against synthesized phosphopeptide derived from human BAD around the phosphorylation site of serine 112 (H-S-SP-Y-P). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| BAD (specific to Phospho-Ser112) detects endogenous levels of BAD only when phosphorylated at serine 112. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction