Learn More
Invitrogen™ BAG1 Monoclonal Antibody (GP3.10G3E2)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA561959
Product Code. 30535721
  • 429.00 € / 100µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The oncogene BCL2 is a membrane protein that blocks a step in a pathway leading to apoptosis or programmed cell death. The protein encoded by this gene binds to BCL2 and is referred to as BCL2-associated athanogene. It enhances the anti-apoptotic effects of BCL2 and represents a link between growth factor receptors and anti-apoptotic mechanisms. At least three protein isoforms are encoded by this mRNA through the use of a non-AUG (CUG) start site, and alternative, downstream, AUG translation initiation sites.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BAG1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q99933
Bag1
E. coli-derived recombinant human Bag-1.
100 μg
Primary
Human
Antibody
IgG1
Immunoprecipitation, Western Blot
GP3.10G3E2
Unconjugated
Bag1
BAG family molecular chaperone regulator 1; Bag1; BAG-1; BCL2 associated athanogene 1; Bcl-2 associating athanogene-1 protein; BCL2-associated athanogene; bcl-2-associated athanogene 1; BCL2-associated athanogene 1; Bcl-2-binding protein; glucocortoid receptor-associated protein RAP46; HAP; Rap46; receptor-associated protein, 46-KD; RP11-326F20.2
Mouse
Protein A/G
RUO
573
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized