Learn More
BAG3 Antibody (3D8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26483-100ul
 View more versions of this product
Product Code. 30029802
  • 533.00 € / 100µL
Description
BAG3 Monoclonal antibody specifically detects BAG3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BAG3
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry
3D8
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307
A synthesized peptide derived from Human BAG3 [UniProt O95817]
100 μL
Apoptosis, Cancer, Cell Biology, Endocrinology
9531
Store at -20 to -70C. Avoid freeze-thaw cycles.