Learn More
Novus Biologicals™ BAG3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30242
Product Code. 30045246
  • 571.00 € / 20µg
Description
BAG3 Polyclonal antibody specifically detects BAG3 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BAG3
Polyclonal
Tris-buffered Saline 0.1% BSA
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307
between 300 and 350
20 μg
Apoptosis, Cancer, Cell Biology, Endocrinology
9531
Store at 4°C. Do not freeze.