missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BAG3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29130
This item is not returnable.
View return policy
Description
BAG3 Polyclonal antibody specifically detects BAG3 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| BAG3 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307 | |
| between 525 and 575 | |
| 20 μg | |
| Apoptosis, Cancer, Cell Biology, Endocrinology | |
| 9531 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction