Learn More
BAG3 Antibody (V65P1E8*D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50112
 View more versions of this product
Product Code. 18693068
  • 351.00 € / 0.10mL
Description
BAG3 Monoclonal antibody specifically detects BAG3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BAG3
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307
Ovalbumin-conjugated synthetic peptide MTDTPGNPAAP
0.1 mL
Apoptosis, Cancer, Cell Biology, Endocrinology
9531
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry
V65P1E8*D2
Unconjugated
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified