Learn More
BAG3 Antibody (V65P1E8*D2), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50112JF646
Product Code. 18776901
  • 437.00 € / 0.10mL
Description
BAG3 Monoclonal antibody specifically detects BAG3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BAG3
Monoclonal
Janelia Fluor 646
BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307
Ovalbumin-conjugated synthetic peptide MTDTPGNPAAP
0.1 mL
Apoptosis, Cancer, Cell Biology, Endocrinology
9531
Store at 4°C in the dark.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry
V65P1E8*D2
50 mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified