missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bags and Liners for HyPerforma™ Single-Use Mixer DS 300, 100L Liner - Bottom drain, non-irradiated

Product Code. 17820657 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 L
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17820657 100 L Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17820657 Supplier Thermo Scientific™ Supplier No. SH30399.02
  • ONLINE EXCLUSIVE PRICE
    1615.00 €
    1319.83 € / Pack of 5
    Save 295.17 €
    18% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The 100L open-top, non-irradiated BioProcess Container (BPC) tank liner with bottom drain is composed of CX3-9 single web, multi-layer plastic film and compatible with Thermo Scientific™ HyPerforma Single-Use Mixer DS 300.

The 100L open-top, non-irradiated BioProcess Container (BPC) tank liner with bottom drain is composed of CX3-9 single web, multi-layer plastic film and compatible with Thermo Scientific™ HyPerforma Single-Use Mixer DS 300. Implement industry-proven BPCs and liners in the overall process of shipping and storing your liquid-liquid and powder-liquid bioprocess mixing applications. BPCs and liners come in open- and closed-top design and are manufactured using our proprietary CX5-14 and CX3-9 film that easily fit in every plastic drum size.

Key Features of HyPerforma Single-Use Mixer DS 300 BPCs and liners:
• All standard BPCs and liners are designed to fit “out-of-the-box“
• Available in volume options of 50, 100, and 200L
• Applicable to open- and closed-top mixing applications
• Powder-liquid and liquid-liquid mixing
• World class CX5-14 and CX3-9 film
• Adaptable to multiple upstream process applications in media and buffer prep

Required Accessories:
• HyPerforma Single-Use Mixer DS 300 docking station
• HyPerforma SIngle-Use Mixer DS 300 drums

Recommended for:
• Powder-liquid or liquid-liquid mixing applications in single use
• High-throughput mixing and storage
• Additional configurations are available to suit your application requirements.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 100 L
Film Description CX3-9
Port Detail Bottom Port: 0.5 in.
Outer Packaging Packages placed in strong cardboard cartons with product and lot identification
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity 100 L
Sterilization Method Non-Irradiated
Tubing Line 1 Bottom Port: 1.27 x 1.91 x 122 cm (0.5 x 0.75 x 304.1 in.) Polycarbonate Quick Connect 12.7 mm (0.5 in.) insert (MPX) and cap
Product Line HyPerforma
Tubing Style Impeller Sleeve with Quick Connect: C-Flex™ Tubing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.