missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAI1 Isoform 3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11566083
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11566083 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11566083 Supplier Invitrogen™ Supplier No. PA131418

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

PA1-31418 has been successfully used in Western blot applications, where a band of approximately 50 kDa is detected. Overnight incubation is recommended. Positive control: HeLa cell lysates. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

BAI 1 is likely to be a potent inhibitor of angiogenesis in brain and may play a significant role as a mediator of the p53 signal in suppression of glioblastoma. It may function in cell adhesion and signal transduction in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAI1 Isoform 3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ADGRB1
Gene Accession No. O14514
Gene Alias ADGRB1; Adhesion G protein-coupled receptor B1; BAI1; Brain-specific angiogenesis inhibitor 1; FLJ41988; GDAIF; Vasculostatin-120; Vasculostatin-40; Vstat120; Vstat40
Gene Symbols ADGRB1
Host Species Goat
Immunogen Peptide with sequence C-SMSSADVEVARF, from the C Terminus of the protein sequence.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 575
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.