missing translation for 'onlineSavingsMsg'
Learn More

Baicalein, MedChemExpress

Product Code. 16489026
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489026 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16489026 Supplier Medchem Express Supplier No. HYN0196
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Baicalein (5,6,7-Trihydroxyflavone) is a xanthine oxidase inhibitor with an IC50 value of 3.12 μM.

Baicalein (5,6,7-Trihydroxyflavone) is a xanthine oxidase inhibitor with an IC50 value of 3.12 μM.

Specifications

Chemical Name or Material Baicalein
CAS 491-67-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10O5
Quantity 100 mg
Synonym 5,6,7-Trihydroxyflavone
Solubility Information DMSO : 50 mg/mL (185.02 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=CC=C2)OC3=CC(O)=C(O)C(O)=C13
Molecular Weight (g/mol) 270.24
Formula Weight 270.24
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.