missing translation for 'onlineSavingsMsg'
Learn More

Baloxavir marboxil, MedChemExpress

Product Code. 16475186
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475186 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16475186 Supplier Medchem Express Supplier No. HY109025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Baloxavir marboxil (S-033188) is a selective inhibitor of influenza cap-dependent endonuclease. Baloxavir marboxil, a potent antiviral agent, shows activity against influenza A and B virus.

Baloxavir marboxil (S-033188) is a selective inhibitor of influenza cap-dependent endonuclease. Baloxavir marboxil, a potent antiviral agent, shows activity against influenza A and B virus.

Specifications

Chemical Name or Material Baloxavir marboxil
Physical Form Solid
CAS 1985606-14-1
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C27H23F2N3O7S
For Use With (Application) COVID-19-anti-virus
Synonym S-033188
Solubility Information DMSO : 33.33 mg/mL (58.32 mM; Need ultrasonic)
SMILES O=C(C=C1)C(OCOC(OC)=O)=C2N1N([C@H]3C4=CC=C(F)C(F)=C4CSC5=CC=CC=C35)[C@@]6([H])N(CCOC6)C2=O
Molecular Weight (g/mol) 571.55
Formula Weight 571.55
Percent Purity 98.94%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.