missing translation for 'onlineSavingsMsg'
Learn More

Balzers chrome layer etching solution, Fluka™

Product Code. 15677030 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 L
200 L
Packaging:
Metal Drum
Plastic Pail
Unit Size:
200 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15677030 25 L Plastic Pail 25 L
15667030 200 L Metal Drum 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15677030 Supplier Honeywell-Fluka Supplier No. 7472725L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Balzers chrome layer etching solution

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Balzers chrome layer etching solution
Color Light Yellow
CAS 16774-21-3, 64-19-7, 7732-18-5
Boiling Point 100°C (1.013 hPa)
Grade Synthesis
Packaging Plastic Pail
pH Acidic
Quantity 25 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.