missing translation for 'onlineSavingsMsg'
Learn More

Bambuterol Monocarbamate Hydrochloride, TRC

Product Code. 30132975
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132975 50 mg 50mg
17970605 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132975 Supplier Toronto Research Chemicals Supplier No. TRCB11751550MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81732-52-7
Molecular Formula C15 H24 N2 O4 . H Cl
Molecular Weight (g/mol) 332.82
Synonym Carbamic acid, dimethyl-, 3-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-5-hydroxyphenyl ester, monohydrochloride (9CI), Bambuterol monocarbamate hydrochloride, 3-[(1RS)-2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-5-hydroxyphenyl dimethylcarbamate hydrochloride
IUPAC Name [3-[2-(tert-butylamino)-1-hydroxyethyl]-5-hydroxyphenyl] N,N-dimethylcarbamate;hydrochloride
SMILES Cl.CN(C)C(=O)Oc1cc(O)cc(c1)C(O)CNC(C)(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H24N2O4.ClH/c1-15(2,3)16-9-13(19)10-6-11(18)8-12(7-10)21-14(20)17(4)5;/h6-8,13,16,18-19H,9H2,1-5H3;1H
Formula Weight 332.1503
Chemical Name or Material Bambuterol Monocarbamate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.