missing translation for 'onlineSavingsMsg'
Learn More

Bamethan Hemisulfate Salt, TRC

Product Code. 30138272
Change view
Click to view available options
Quantity:
100 mg
5 g
Unit Size:
100mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138272 5 g 5g
30005031 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138272 Supplier Toronto Research Chemicals Supplier No. TRCB1179005G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5716-20-1
Molecular Formula 2 C12 H19 N O2 . H2 O4 S
Molecular Weight (g/mol) 516.648
Synonym Benzenemethanol, α-[(butylamino)methyl]-4-hydroxy-, sulfate (2:1) (salt) (9CI), Benzyl alcohol, α-[(butylamino)methyl]-p-hydroxy-, sulfate (2:1) (salt) (8CI), (±)-1-(4-Hydroxyphenyl)-1-hydroxy-2-butylaminoethane sulfate, 1-(4-Hydroxyphenyl)-2-butylaminoethanol-sulfate, 2-n-Butylamino-1-(p-hydroxyphenyl)ethanol sulfate, Bametan sulfate, Bamethan sulfate, Bascurat, Bupatol, Butedrin, Butibatol, Butylnorsympatol, Cyclate, Eclern, Garmian, Periphetol, Rotesar, Vasculat, Vasculit, Vaskulat, α-[(Butylamino)methyl]-p-hydroxybenzyl alcohol sulfate (2:1), Benzenemethanol, α-[(butylamino)methyl]-4-hydroxy-, sulfate (2:1), Bamethan Sulfate
IUPAC Name 4-[2-(butylamino)-1-hydroxyethyl]phenol;sulfuric acid
SMILES CCCCNCC(O)c1ccc(O)cc1.CCCCNCC(O)c2ccc(O)cc2.OS(=O)(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/2C12H19NO2.H2O4S/c2*1-2-3-8-13-9-12(15)10-4-6-11(14)7-5-10;1-5(2,3)4/h2*4-7,12-15H,2-3,8-9H2,1H3;(H2,1,2,3,4)
Formula Weight 516.2505
Chemical Name or Material Bamethan Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.