missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAP28 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92105-0.1ml
This item is not returnable.
View return policy
Description
BAP28 Polyclonal antibody specifically detects BAP28 in Human samples. It is validated for Western Blot
Specifications
| BAP28 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BAP28HEAT repeat-containing protein 1, EC 4.1.1.31, EC 6.3.4.4, FLJ10359, HEAT repeat containing 1, MGC72083, Protein BAP28 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human HEATR1 (NP_060542.4). MTSLAQQLQRLALPQSDASLLSRDEVASLLFDPKEAATIDRDTAFAIGCTGLEELLGIDPSFEQFEAPLFSQLAKTLERSVQTKAVNKQLDENISLFLIHLSPYFLLKPAQKCLEWLIHR | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 55127 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction