missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride, 0.5N Standardized Solution

Product Code. 11323687
Change view
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
Product Code. Quantity unitSize
11323687 4 L 4 L
11333687 1 L 1 L
2 options
This item is not returnable. View return policy

Product Code. 11323687

Brand: Thermo Scientific Alfa Aesar 035583.K7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10361-37-2 | BaCl2 | 208.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-37-2
Molecular Formula BaCl2
Molecular Weight (g/mol) 208.23
MDL Number MFCD00003445
InChI Key WDIHJSXYQDMJHN-UHFFFAOYSA-L
Synonym barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
PubChem CID 25204
ChEBI CHEBI:63317
IUPAC Name barium(2+);dichloride
SMILES [Cl-].[Cl-].[Ba++]

Specifications

Color Colorless
Concentration 0.5N
Formula Weight 208.25
Odor Odorless
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Barium chloride, Standardized Solution

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.