missing translation for 'onlineSavingsMsg'
Learn More

Barium Standard: Ba @ 1000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 17971896
Change view
Click to view available options
Quantity:
50 g
200 g
Unit Size:
200g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971896 50 g 50g
17901856 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17971896 Supplier VHG Labs Supplier No. VHGOBA100050G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Barium Standard: Ba @ 1000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Barium
CAS 7440-39-3
Methods ICP/XRF/RDE
Analyte or Component Names Barium
Shipping Condition Room Temperature
Molecular Formula Ba
Certifications/Compliance ISO 17034
Synonym Bario, Baryum
InChI Formula InChI=1S/Ba
SMILES [Ba]
IUPAC Name barium
Solvent or Matrix 75 cSt Mineral Oil
Molecular Weight (g/mol) 137.33
Quantity 50 g
Formula Weight 137.905 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.