missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Basal Medium Eagle (Earle's)

Product Code. 31908462 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
10 x 500 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908462 10 x 500 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908462 Supplier Sartorius Supplier No. 010155A
 Restricted Item
Estimated Shipment: 25-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy

BME is a widely used synthetic basal medium for supporting the growth of many different mammalian cells.

  • BME was originally developed by Harry Eagle for HeLa cells and mouse fibroblasts after determining the minimum requirements for cell growth in vitro.
  • BME has since been used for other human cell lines, including WI-38 and MRC-5.
  • BME contains eight B vitamins, ten essential amino acids, plus cystine, tyrosine, and glutamine.
  • Several modifications of BME have been developed, including Minimal Essential Medium (MEM), Glasgow's Medium, and Dulbecco's Modified Eagle's Medium (DMEM).
  • This BME formulation contains Earle's salts for use in a CO2 incubator.
  • Modified as follows:
    • With Earle's Salts Base
    • With Phenol Red
    • Without L-Glutamine
    • Without Sodium Bicarbonate

Specifications

Form Liquid
Product Type MEM Cell Culture Media
Quantity 10 x 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.