missing translation for 'onlineSavingsMsg'
Learn More

Basic fuchsin hydrochloride, 99%, For HPLC analysis, MP Biomedicals™

Product Code. 11423301 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423301 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11423301 Supplier MP Biomedicals Supplier No. 0215042325
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 569-61-9
Color Green
Assay Percent Range 99%
Molecular Formula C19H18ClN3
Quantity 25 g
Synonym basic red 9, pararosaniline hydrochloride, parafuchsin, pararosaniline chloride, parafuchsine, p-fuchsin, basic fuchsin, ci basic red 9, c.i. basic red 9, parafuksin
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
SMILES C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
Molecular Weight (g/mol) 323.824
PubChem CID 11292
ChEBI CHEBI:87663
Formula Weight 323.83
Physical Form Crystalline Powder
Chemical Name or Material Basic fuchsin hydrochloride, For HPLC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.