missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BATF3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41296-0.025mg
This item is not returnable.
View return policy
Description
BATF3 Polyclonal specifically detects BATF3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| BATF3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| BATF3 | |
| Antibody was raised against a 16 amino acid peptide near the amino terminus of human BATF3. The immunogen is located within the first 50 amino acids of BATF3. Amino Acid Squence: APGNQPQPQPQQQSPE | |
| Affinity Purified | |
| RUO | |
| 55509 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5 μg/mL, ELISA, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin, Immunofluorescence | |
| basic leucine zipper transcription factor, ATF-like 3, basic leucine zipper transcriptional factor ATF-like 3, B-ATF-3, FLJ36352, FLJ37535,21 kDa small nuclear factor isolated from T-cells, JDP1, Jun dimerization protein 1, Jun dimerization protein p21SNFT, JUNDM1, SNFT21-kD small nuclear factor isolated from T cells | |
| Rabbit | |
| 17 kDa | |
| 0.025 mg | |
| Primary | |
| BATF3 antibody is predicted to have no cross-reactivity to other BATF family members. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction