missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BATF3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17186883
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17186883 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17186883 Supplier Invitrogen™ Supplier No. PA5107080

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Yeast: 83%.

BATF3, also known as SNFT, belongs to the basic leucine zipper transcriptional family and functions as a transcriptional repressor when heterodimerizing with Jun and can repress the transcription of IL-2 and MMP-1. BATF3 is required for the development of CD8alpha+ classical dendritic cells (DCs) and the related CD103+ DCs that cross-present antigens to CD8 T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BATF3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene BATF3
Gene Accession No. Q9NR55
Gene Alias 21 kDa small nuclear factor isolated from T-cells; 21-kD small nuclear factor isolated from T cells; basic leucine zipper ATF-like transcription factor 3; basic leucine zipper transcription factor, ATF-like 3; basic leucine zipper transcriptional factor ATF-like 3; Batf3; B-ATF-3; JDP1; JDP-1; Jun dimerization protein 1; Jun dimerization protein p21SNFT; JUNDM1; SNFT
Gene Symbols BATF3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human BATF3 (aa 1-51).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55509
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.