missing translation for 'onlineSavingsMsg'
Learn More

Bathocuproin, 98%

Product Code. 11459877
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459877 1 g 1g
11469877 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459877 Supplier Thermo Scientific Alfa Aesar Supplier No. B22841.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

CAS: 4733-39-5 | C26H20N2 | 360.46 g/mol

It is used as a reagent for the determination of copper. It acts as an exciton blocking barrier which prohibits excitons diffusion process towards the Al electrode otherwise being quenched. It is the most commonly used buffer layer between acceptor and cathode layer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent for the determination of copper. It acts as an exciton blocking barrier which prohibits excitons diffusion process towards the Al electrode otherwise being quenched. It is the most commonly used buffer layer between acceptor and cathode layer.

Solubility
Soluble in methanol (36mg/100ml), ethanol, 2-propanol, bezene, and acetone. Partly miscible in water.

Notes
Store in cool, dry conditions in a sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 4733-39-5
Molecular Formula C26H20N2
Molecular Weight (g/mol) 360.46
MDL Number MFCD00004972
InChI Key STTGYIUESPWXOW-UHFFFAOYSA-N
Synonym bathocuproine, bathocuproin, 1,10-phenanthroline, 2,9-dimethyl-4,7-diphenyl, unii-9thp2v94fx, 9thp2v94fx, 2,9-dimethyl-4,7-diphenylpyridino 3,2-h quinoline, pubchem19657, bathocuproin 1g, bidd:gt0563
PubChem CID 65149
IUPAC Name 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline
SMILES CC1=CC(C2=CC=CC=C2)=C2C=CC3=C(C=C(C)N=C3C2=N1)C1=CC=CC=C1

Tekniske data

Melting Point 279°C to 285°C
Quantity 1 g
Beilstein 306714
Solubility Information Soluble in methanol (36mg/100ml),ethanol,2-propanol,bezene,and acetone. Partly miscible in water.
Formula Weight 360.46
Percent Purity 98%
Chemical Name or Material Bathocuproin

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.