Learn More
CleanSpace Technology™ Battery Charger
Product Code. 15970092 Shop All CleanSpace Technology Products
Change view
Click to view available options
Product Type:
Charger
Unit Size:
Each
Product Code. Product Type Unit Size
15970092 Charger Each
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15970092 Supplier CleanSpace Technology™ Supplier No. PAF1101
  • ONLINE EXCLUSIVE PRICE
    98.41 €
    76.00 € / Pack of 1
    Save 22.41 €
    22% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
The CleanSpace Battery Charger is specifically designed to re-charge the internal lithium polymer battery of the CleanSpace Power Units. The battery charger has two leads and interchangeable plugs for use in different countries.
  • Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
  • Multinational interchangeable plugs
  • Dual leads for charging two PAPR power units simultaneously
Specifications
Product Type Charger
For Use With (Equipment) PAF-0034, PAF-0060, PAF-0070