missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bax Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
227.00 € - 319.00 €
Specifications
| Antigen | Bax |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18785403
|
Novus Biologicals
NB100-56096SS |
0.025 mL |
227.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18729843
|
Novus Biologicals
NB100-56096 |
0.05 mL |
319.00 €
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bax Polyclonal specifically detects Bax in Human, Mouse, Rat, Canine, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Bax | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
| Whole antisera with 0.05% Sodium Azide | |
| apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4 | |
| BAX | |
| IgG | |
| Unpurified | |
| Bax |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Canine, Gerbil | |
| Q07813 | |
| 581 | |
| A synthetic peptide corresponding to amino acids 43-60 (PELTLEQPPQDASTKKLS) of mouse Bax alpha was used as immunogen, GenBank no. NP_031553.1. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| 21 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title