missing translation for 'onlineSavingsMsg'
Learn More

BAY 41-2272, MedChemExpress

Product Code. 16465846
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465846 100 mg 100mg
16463296 25 mg 25mg
16457847 5 mg 5mg
16435128 10 mg 10mg
16401367 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16465846 Supplier Medchem Express Supplier No. HY12376
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BAY 41-2272 is a soluble guanylate cyclases (sGC) activator.

BAY 41-2272 is a soluble guanylate cyclases (sGC) activator.

Specifications

Chemical Name or Material BAY 41-2272
CAS 256376-24-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H17FN6
Quantity 100 mg
Solubility Information DMSO : 17.5 mg/mL (48.56 mM; Need ultrasonic)
SMILES NC1=NC(C2=NN(CC3=CC=CC=C3F)C4=NC=CC=C42)=NC=C1C5CC5
Molecular Weight (g/mol) 360.39
Formula Weight 360.39
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.