Learn More
Draeger Safety Bayonet A1B1E1K1HgP3 R D Combination Cartridge
Description
- Successfully passed a dolomite test
- Can be used for protection in a wide range of applications
- Easy to use and attach
- Filters are secured in a top-down position
- Maximum protection provided due to the individual packaging
- Weight is 140g per piece
- CE-certified (EN 14387, EN 143:2000/A1:2006)
- Australian Standard AS /NZS 1716 certified (SAI Global)
Specifications
Specifications
| Filter Type | A1B1E1K1 Hg P3 R D |
| Quantity | 1 Ea. |