missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: CHROMagar™ Candida
Description
- BBL™ CHROMagar™ Candida Medium is a selective and differential medium for the isolation of fungi.
- With the inclusion of chromogenic substrates in the medium, the colonies of C. albicans, C. tropicalis and C. krusei produce different colors, thus allowing the direct detection of these yeast species on the isolation plate.
- Colonies of C. albicans appear light to medium green.
- C. tropicalis colonies appear blue-greenish to metallic-blue.
- C. krusei colonies appear light rose with a whitish border.
- Other yeast species may develop either their natural color (cream) or appear rose or light to dark mauve.
- An additional advantage of the medium is the easy detection of mixed yeast cultures due to their colony appearance in different colors.
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Beige |
| Country of Origin | US |
| Form | Liquid |
| Media Type | CHROMagar™ Candida |
| Formulation | Chromopeptone 10.0 g/L, Glucose 20.0 g/L, Chromogen Mix 2.0 g/L, Chloramphenicol 0.5 g/L, Agar 15.0 g/L |
| For Use With (Application) | Yeast and Filamentous Fungi and Differentiation of Candida Strains. |
| No. per Pack | 20 |
| Packaging Type | Plate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?