missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92569-0.1ml
This item is not returnable.
View return policy
Description
BBS10 Polyclonal antibody specifically detects BBS10 in Human, Mouse samples. It is validated for Western Blot
Specifications
| BBS10 | |
| Polyclonal | |
| Western Blot 1:200-1:3000 | |
| Bardet-Biedl syndrome 10, Bardet-Biedl syndrome 10 protein, FLJ23560C12orf58chromosome 12 open reading frame 58 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 474-723 of human BBS10 (NP_078961.3). AENKDALEKTQTYLKVHSNLVIPDVELETYIPYSTPTLTPTDTFQTVETLTCLSLERNRLTDYYEPLLKNNSTAYSTRGNRIEISYENLQVTNITRKGSMLPVSCKLPNMGTSQSYLSSSMPAGCVLPVGGNFEILLHYYLLNYAKKCHQSEETMVSMIIANALLGIPKVLYKSKTGKYSFPHTYIRAVHALQTNQPLVSSQTGLESVMGKYQLLTSVLQCLTKILTIDMVITVKRHPQKVHNQDSEDEL | |
| 0.1 mL | |
| Vision | |
| 79738 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction