missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBSome Interacting Protein 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30510
This item is not returnable.
View return policy
Description
BBSome Interacting Protein 1 Polyclonal specifically detects BBSome Interacting Protein 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BBSome Interacting Protein 1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| A8MTZ0 | |
| BBIP1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EVKSMFREVLPKQGPLFVEDIMTMVLCKPKLLPLKSLTLEKLEKMHQAAQNTIRQQEMAEKDQRQ | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| bA348N5.3, BBIP1, BBIP10, BBSome-Interacting Protein 1, BBSome-Interacting Protein Of 10 KDa, NCRNA00081, Non-Protein Coding RNA 81, UPF0604 Protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 92482 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction