Learn More
BCAT2 Mouse, anti-Human, Clone: 1A5A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68586-1-IG-20UL
This item is not returnable.
View return policy
Description
The BCAT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BCAT2. This antibody recognizes human, mouse antigen. The BCAT2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BCAT2 | |
| 1A5A5 | |
| BCAT2, ECA40, Placental protein 18, PP18 | |
| 20 μL | |
| 587 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG2a |