Learn More
Bcl-2 Antibody (CL7935) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88910AF647
Product Code. 30307025
  • 759.00 € / 0.10mL
Description
Bcl-2 Monoclonal antibody specifically detects Bcl-2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bcl-2
Monoclonal
Alexa Fluor 647
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2
This antibody was developed using a synthetic peptide derived from P10415, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis, Autophagy, Biologically Active Proteins, Cancer, Cellular Markers, Mitochondrial Mediated Pathway, Mitophagy, Oncogenes, Phospho Specific, Tumor Suppressors
596
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL7935
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified