missing translation for 'onlineSavingsMsg'
Learn More

Bcl-6 Antibody (CL14611) [Alexa Fluor™ 700], Novus Biologicals™

Product Code. 30297053 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30297053 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30297053

Brand: Novus Biologicals NBP326872AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Bcl-6 Monoclonal antibody specifically detects Bcl-6 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-6
Applications Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL14611
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias B-cell CLL/lymphoma 6, B-cell lymphoma 6 protein transcript, BCL-5, BCL5B-cell lymphoma 5 protein, BCL-6, BCL6A, cys-his2 zinc finger transcription factor, LAZ3ZBTB27B-cell lymphoma 6 protein, lymphoma-associated zinc finger gene on chromosome 3, Protein LAZ-3, Zinc finger and BTB domain-containing protein 27, Zinc finger protein 51ZNF51, zinc finger transcription factor BCL6S
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from P41182, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 604
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.