missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Bcl-6 Antibody (CL14612), Novus Biologicals™
Mouse Monoclonal Antibody
317.00 € - 501.00 €
Specifications
| Antigen | Bcl-6 |
|---|---|
| Clone | CL14612 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL |
| Applications | Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043118
|
Novus Biologicals™
NBP3-26873-25ul |
25 μL |
317.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043989
|
Novus Biologicals™
NBP3-26873-100ul |
100 μL |
501.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bcl-6 Monoclonal antibody specifically detects Bcl-6 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry/ ImmunofluorescenceSpecifications
| Bcl-6 | |
| 1 mg/mL | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Tumor Suppressors | |
| PBS, pH 7.2, 40% glycerol | |
| 604 | |
| IgG2a | |
| Protein A purified |
| CL14612 | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B-cell CLL/lymphoma 6, B-cell lymphoma 6 protein transcript, BCL-5, BCL5B-cell lymphoma 5 protein, BCL-6, BCL6A, cys-his2 zinc finger transcription factor, LAZ3ZBTB27B-cell lymphoma 6 protein, lymphoma-associated zinc finger gene on chromosome 3, Protein LAZ-3, Zinc finger and BTB domain-containing protein 27, Zinc finger protein 51ZNF51, zinc finger transcription factor BCL6S | |
| This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of P41182. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title