missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bcl-B Polyclonal Antibody

Product Code. 12888392
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12888392 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12888392 Supplier Invitrogen™ Supplier No. PA520234
  • ONLINE EXCLUSIVE PRICE
    530.00 €
    376.30 € / 100µg
    Save 153.70 €
    29% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Despite its predicted molecular weight, Bcl-B often migrates at higher molecular weights, presumably due to post-translational modifications. A suggested positive control is Jurkat cell lysate. PA5-20234 can be used with blocking peptide PEP-0353.

The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The protein encoded by this gene contains conserved BH4, BH1 and BH2 domains. This protein can interact with other members of BCL-2 protein family including BCL2, BCL2L1/BCL-X(L), and BAX. Overexpression of this gene has been shown to suppress cell apoptosis possibly through the prevention of cytochrome C release from the mitochondria, and thus activating caspase-3 activation. The mouse counterpart of this protein is found to interact with Apaf1 and forms a protein complex with Caspase 9, which suggests the involvement of this protein in APAF1 and CASPASE 9 related apoptotic pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene BCL2L10
Gene Accession No. Q9HD36
Gene Alias AA420380; anti-apoptotic protein Boo; Anti-apoptotic protein NrH; apoptosis regulator Bcl-B; AU023065; Bcl-2 homolog Diva; BCL2 like 10; BCL2L10; bcl2-L-10; Bcl2-like 10; BCL2-like 10 (apoptosis facilitator); bcl-2-like protein 10; BCLB; BCL-B; Boo; C85687; death inducer binding to vBcl-2 and Apaf-1; Diva
Gene Symbols BCL2L10
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the amino terminus of human Bcl-B.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10017
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.