missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92041-0.1ml
This item is not returnable.
View return policy
Description
Bcl3 Polyclonal antibody specifically detects Bcl3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Bcl3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| B-cell CLL/lymphoma 3, B-cell leukemia/lymphoma 3, B-cell lymphoma 3 protein, B-cell lymphoma 3-encoded protein, BCL-3, BCL4, chronic lymphatic leukemia protein, D19S37, Proto-oncogene BCL3 | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human BCL3 (NP_005169.2). ANVNAQMYSGSSALHSASGRGLLPLVRTLVRSGADSSLKNCHNDTPLMVARSRRVIDILRGKATRPASTSQPDPSPDRSANTSPESSSRLSSNGLLSASPS | |
| 0.1 mL | |
| Adaptive Immunity, Cell Cycle and Replication, Chromatin Research, Immunology, Signal Transduction, Transcription Factors and Regulators | |
| 602 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction