missing translation for 'onlineSavingsMsg'
Learn More

BCL7B Antibody (OTI8E10), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-44816-100ul

 View more versions of this product

Product Code. 30306356

  • 550.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

BCL7B Monoclonal antibody specifically detects BCL7B in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BCL7B
Monoclonal
Unconjugated
PBS (pH 7.3) 1% BSA, 50% glycerol
Mouse
>95%
RUO
9275
Store at -20 degrees C. Avoid freeze/thaw cycles.
Purified
Western Blot
OTI8E10
Western Blot 1:2000
Allergen Hom s 3, B-cell CLL/lymphoma 7 protein family member B, B-cell CLL/lymphoma 7B
Full length human recombiant protein of human BCL7B (NP_001698) produced in E.coli.
100 μL
Primary
Human, Mouse
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.