Learn More
Novus Biologicals™ BCMA/TNFRSF17 Antibody (belantamab), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28602-1mg
 View more versions of this product
Product Code. 30048025
  • 920.00 € / 1mg
Description
BCMA/TNFRSF17 Monoclonal antibody specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BCMA/TNFRSF17
Monoclonal
Unconjugated
Q02223
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
belantamab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17
TNFRSF17/ BCMA/ CD269
1 mg
Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Signal Transduction
608
Human
Purified