missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCS1L Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92916-0.02ml
This item is not returnable.
View return policy
Description
BCS1L Polyclonal antibody specifically detects BCS1L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| BCS1L | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| BCS, BCS1, BCS1 (yeast homolog)-like, BCS1-like (S. cerevisiae), BCS1-like (yeast), BCS1-like protein, BJSGRACILE, FLNMS, h-BCS, h-BCS1, mitochondrial chaperone BCS1, mitochondrial complex III assembly, PTD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 33-240 of human BCS1L (NP_004319.1). RKGVQLGLVAFRRHYMITLEVPARDRSYAWLLSWLTRHSTRTQHLSVETSYLQHESGRISTKFEFVPSPGNHFIWYRGKWIRVERSREMQMIDLQTGTPWESVTFTALGTDRKVFFNILEEARELALQQEEGKTVMYTAVGSEWRPFGYPRRRRPLNSVVLQQGLADRIVRDVQEFIDNPKWYTDRGIPYRRGYLLYGPPGCGKSSFI | |
| 0.02 mL | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 617 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering