missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Difco™ Casamino Acids, vitamin assay
Excellent supplement for media formulations where nitrogen requirements are minimal, including diphtheria toxin production. Gibco™ Bacto™ Casamino Acids is an animal-origin, acid hydrolysis of casein having low sodium chloride and iron concentrations.
139.00 € - 586.00 €
Specifications
| Certifications/Compliance | AOAC, BAM, COMPF, SMWW, USDA, USP, ISO/DIS, 11014 |
|---|---|
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Product Type | Casamino Acids |
| For Use With (Application) | Vitamin assay |
| Packaging Type | Bottle |
| Product Code | Brand | For Use With (Application) | Quantity | dimGrade | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | For Use With (Application) | Quantity | dimGrade | Price | Quantity & Availability | |||
|
16219861
|
Gibco™
228820 |
Vitamin assay | 100 g |
139.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
16229861
|
Gibco™
228830 |
Vitamin assay | 500 g |
586.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Prepared according to the method described by Mueller and Miller
- Supplemented with growth factors, cystine, maltose and iron and inorganic salts
- Successful in fermentation for Pneumococcal, Diptheria, Pertussis, Tetanus and Zoster vaccines
- Support the growth of Bordetella, Corynebacterium, Pseudomonas, Haemophilus and Escherichia
- Dehydrated format with Peptones / Hydrolysates
- DNR (Latex) free
Specifications
| AOAC, BAM, COMPF, SMWW, USDA, USP, ISO/DIS, 11014 | |
| Casamino Acids | |
| Bottle |
| Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. | |
| Vitamin assay | |
| Difco™ |
Safety and Handling
missing translation for 'shelfLife' : 1825 Days
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title