missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Mycoflask™ Urea Agar Slants, Complete (Christensen) Prepared Media

Product Code. 16411531
Change view
Click to view available options
Quantity:
10/Pk.
100 Pack
Unit Size:
Pack of 10
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16411531 100 Pack Pack of 100
17342321 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16411531 Supplier BD Supplier No. 221097
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for the differentiation of organisms, especially the Enterobacteriaceae, on the basis of urease production

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM, CCAM, ISO, USDA
Color Yellow
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Urea Agar
Formulation Pancreatic digest of Gelatin 1g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Potassium Phosphate 2g/L, Urea 20g/L, Phenol red 0.012g/L, Agar 15g/L
For Use With (Application) For differentiation of organisms, especially Enterobacteriaceae, on basis of Urease production
Packaging Type Glass Tube with Cap
Quantity 100 Pack

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.