Learn More
BD BBL™ Prepared Plated Media: BCYE Agar (for Legionella)
Product Code. 17284353
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. Quantity unitSize
17284353 10/Pk. Pack of 10
1 options
Product Code. 17284353 Supplier BD Supplier No. 221808
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For primary isolation and cultivation of Legionella pneumophila and other Legionella species from environmental samples and clinical specimens
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO/DIS, 11014
Color Black
Form Liquid
Manufacturing Origin United States
Media Type BCYE Agar (for Legionella)
Formulation Yeast extract 10g/L, L-Cysteine HCl 0.4g/L, Ferric Pyrophosphate 0.25g/L, ACES Buffer 10g/L, Charcoal, Activated 2g/L, Alpha-Ketoglutarate 1g/L, Agar 15g/L
For Use With (Application) Primary isolation and cultivation of Legionella pneumophila and other Legionella species from environmental samples and clinical specimens
Packaging Type Plate
Quantity 10/Pk.